Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACCCTTTGAGACTACTAATCTG[G/T]TGTCCATTTATATAATTTTATTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614657 | ||||||||||||||||||||
Literature Links: |
AMOTL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMOTL1 - angiomotin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301007.1 | Intron | NP_001287936.1 | ||||
NM_130847.2 | Intron | NP_570899.1 | ||||
XM_005273798.4 | Intron | XP_005273855.1 | ||||
XM_005273801.4 | Intron | XP_005273858.1 | ||||
XM_006718772.3 | Intron | XP_006718835.1 | ||||
XM_011542626.2 | Intron | XP_011540928.1 | ||||
XM_011542628.2 | Intron | XP_011540930.1 |
LOC105369438 - uncharacterized LOC105369438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |