Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGATGAGAGGAGTTTTTCATGG[A/G]AGGTGGTGGGTTGCCATGGATTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609431 | ||||||||||||||||||||
Literature Links: |
MBIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBIP - MAP3K12 binding inhibitory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144891.1 | 945 | Silent Mutation | CTC,CTT | L,L 342 | NP_001138363.1 | |
NM_001308110.1 | 945 | UTR 3 | NP_001295039.1 | |||
NM_016586.2 | 945 | Silent Mutation | CTC,CTT | L,L 343 | NP_057670.2 | |
XM_005267754.3 | 945 | Silent Mutation | CTC,CTT | L,L 321 | XP_005267811.1 | |
XM_005267756.4 | 945 | UTR 3 | XP_005267813.1 | |||
XM_017021367.1 | 945 | UTR 3 | XP_016876856.1 |