Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAATTGTTGAAGTCAGCATTTCC[A/G]GTGAAACTAAAGCTTTTGATGCAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300420 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PJA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PJA1 - praja ring finger ubiquitin ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032396.2 | 2291 | UTR 3 | NP_001027568.1 | |||
NM_022368.4 | 2291 | UTR 3 | NP_071763.2 | |||
NM_145119.3 | 2291 | UTR 3 | NP_660095.1 | |||
XM_005262292.1 | 2291 | Intron | XP_005262349.1 | |||
XM_011531011.2 | 2291 | Intron | XP_011529313.1 |
Set Membership: |
HapMap |