Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATTTATGTCTTCTTATTTCATG[C/T]GAATGCATTTCTTCATAGAATAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616417 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRL3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRL3 - adhesion G protein-coupled receptor L3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322246.1 | Intron | NP_001309175.1 | ||||
NM_001322402.1 | Intron | NP_001309331.1 | ||||
NM_015236.5 | Intron | NP_056051.2 | ||||
XM_011531788.2 | Intron | XP_011530090.1 | ||||
XM_011531791.2 | Intron | XP_011530093.1 | ||||
XM_017007929.1 | Intron | XP_016863418.1 | ||||
XM_017007930.1 | Intron | XP_016863419.1 | ||||
XM_017007931.1 | Intron | XP_016863420.1 | ||||
XM_017007932.1 | Intron | XP_016863421.1 | ||||
XM_017007933.1 | Intron | XP_016863422.1 | ||||
XM_017007934.1 | Intron | XP_016863423.1 | ||||
XM_017007935.1 | Intron | XP_016863424.1 | ||||
XM_017007936.1 | Intron | XP_016863425.1 | ||||
XM_017007937.1 | Intron | XP_016863426.1 | ||||
XM_017007938.1 | Intron | XP_016863427.1 | ||||
XM_017007939.1 | Intron | XP_016863428.1 | ||||
XM_017007940.1 | Intron | XP_016863429.1 | ||||
XM_017007941.1 | Intron | XP_016863430.1 | ||||
XM_017007942.1 | Intron | XP_016863431.1 |
Set Membership: |
HapMap |