Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATCTGACTGGATCGTTAGTTTT[A/C]TGAGGTATACCCCTACTTTACCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614997 | ||||||||||||||||||||
Literature Links: |
GATAD2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GATAD2A - GATA zinc finger domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300946.1 | Intron | NP_001287875.1 | ||||
NM_017660.3 | Intron | NP_060130.3 | ||||
XM_011528105.1 | Intron | XP_011526407.1 | ||||
XM_017026901.1 | Intron | XP_016882390.1 | ||||
XM_017026902.1 | Intron | XP_016882391.1 | ||||
XM_017026903.1 | Intron | XP_016882392.1 | ||||
XM_017026904.1 | Intron | XP_016882393.1 | ||||
XM_017026905.1 | Intron | XP_016882394.1 | ||||
XM_017026906.1 | Intron | XP_016882395.1 | ||||
XM_017026907.1 | Intron | XP_016882396.1 | ||||
XM_017026908.1 | Intron | XP_016882397.1 | ||||
XM_017026909.1 | Intron | XP_016882398.1 | ||||
XM_017026910.1 | Intron | XP_016882399.1 | ||||
XM_017026911.1 | Intron | XP_016882400.1 |