Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAACCCACAGGTTTTTCCTGG[A/G]CCAAGTCTTCCTTTGCTGCCACTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604252 MIM: 614263 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BACE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BACE1 - beta-secretase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207048.1 | 5554 | UTR 3 | NP_001193977.1 | |||
NM_001207049.1 | 5554 | UTR 3 | NP_001193978.1 | |||
NM_012104.4 | 5554 | UTR 3 | NP_036236.1 | |||
NM_138971.3 | 5554 | UTR 3 | NP_620427.1 | |||
NM_138972.3 | 5554 | UTR 3 | NP_620428.1 | |||
NM_138973.3 | 5554 | UTR 3 | NP_620429.1 |
BACE1-AS - BACE1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF214 - ring finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077239.1 | 5554 | Intron | NP_001070707.1 | |||
NM_001278249.1 | 5554 | Intron | NP_001265178.1 | |||
NM_207343.3 | 5554 | Intron | NP_997226.2 |
Set Membership: |
HapMap |