Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAAGTAGAAGAGGAATGTGAGG[A/C]CAAATTTAAAGTAGAAAATGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KA5 - ribosomal protein S6 kinase A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322227.1 | 3372 | UTR 3 | NP_001309156.1 | |||
NM_001322228.1 | 3372 | UTR 3 | NP_001309157.1 | |||
NM_001322229.1 | 3372 | UTR 3 | NP_001309158.1 | |||
NM_001322230.1 | 3372 | UTR 3 | NP_001309159.1 | |||
NM_001322231.1 | 3372 | UTR 3 | NP_001309160.1 | |||
NM_001322232.1 | 3372 | UTR 3 | NP_001309161.1 | |||
NM_001322233.1 | 3372 | UTR 3 | NP_001309162.1 | |||
NM_001322234.1 | 3372 | UTR 3 | NP_001309163.1 | |||
NM_001322235.1 | 3372 | UTR 3 | NP_001309164.1 | |||
NM_001322236.1 | 3372 | UTR 3 | NP_001309165.1 | |||
NM_001322237.1 | 3372 | UTR 3 | NP_001309166.1 | |||
NM_001322238.1 | 3372 | UTR 3 | NP_001309167.1 | |||
NM_004755.3 | 3372 | UTR 3 | NP_004746.2 | |||
NM_182398.2 | 3372 | Intron | NP_872198.1 | |||
XM_017021786.1 | 3372 | Intron | XP_016877275.1 |