Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCTTTTGGGATGATCACATGG[C/T]GAGAATCTGCTTTGAAGATATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608205 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MECR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MECR - mitochondrial trans-2-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024732.2 | 1245 | Missense Mutation | ACC,GCC | T,A 296 | NP_001019903.2 | |
NM_016011.3 | 1245 | Missense Mutation | ACC,GCC | T,A 372 | NP_057095.3 | |
XM_005245885.2 | 1245 | Missense Mutation | ACC,GCC | T,A 400 | XP_005245942.1 | |
XM_005245887.2 | 1245 | Missense Mutation | ACC,GCC | T,A 296 | XP_005245944.1 | |
XM_011541539.1 | 1245 | Missense Mutation | ACC,GCC | T,A 458 | XP_011539841.1 | |
XM_011541540.1 | 1245 | Missense Mutation | ACC,GCC | T,A 430 | XP_011539842.1 | |
XM_011541541.1 | 1245 | UTR 3 | XP_011539843.1 | |||
XM_011541543.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539845.1 | |
XM_011541545.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539847.1 | |
XM_011541546.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539848.1 | |
XM_011541547.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539849.1 | |
XM_011541549.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539851.1 | |
XM_011541550.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539852.1 | |
XM_011541553.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539855.1 | |
XM_011541554.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_011539856.1 | |
XM_017001411.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_016856900.1 | |
XM_017001412.1 | 1245 | Missense Mutation | ACC,GCC | T,A 354 | XP_016856901.1 | |
XM_017001413.1 | 1245 | Missense Mutation | ACC,GCC | T,A 296 | XP_016856902.1 | |
XM_017001414.1 | 1245 | Missense Mutation | ACC,GCC | T,A 296 | XP_016856903.1 | |
XM_017001415.1 | 1245 | Missense Mutation | ACC,GCC | T,A 296 | XP_016856904.1 | |
XM_017001416.1 | 1245 | UTR 3 | XP_016856905.1 |