Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACAGCCAGGCAGTCTCCATGG[C/T]AGTGGAGGCTTCTGAATTTAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF1A - kinesin family member 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244008.1 | 7692 | UTR 3 | NP_001230937.1 | |||
NM_001320705.1 | 7692 | UTR 3 | NP_001307634.1 | |||
XM_005247022.1 | 7692 | UTR 3 | XP_005247079.1 | |||
XM_005247024.1 | 7692 | UTR 3 | XP_005247081.1 | |||
XM_005247027.1 | 7692 | UTR 3 | XP_005247084.1 | |||
XM_005247028.2 | 7692 | UTR 3 | XP_005247085.1 | |||
XM_006712605.1 | 7692 | UTR 3 | XP_006712668.1 | |||
XM_011511364.1 | 7692 | UTR 3 | XP_011509666.1 | |||
XM_011511365.1 | 7692 | UTR 3 | XP_011509667.1 | |||
XM_011511366.2 | 7692 | UTR 3 | XP_011509668.1 | |||
XM_011511367.1 | 7692 | UTR 3 | XP_011509669.1 | |||
XM_017004383.1 | 7692 | UTR 3 | XP_016859872.1 | |||
XM_017004384.1 | 7692 | UTR 3 | XP_016859873.1 | |||
XM_017004385.1 | 7692 | UTR 3 | XP_016859874.1 | |||
XM_017004386.1 | 7692 | UTR 3 | XP_016859875.1 | |||
XM_017004387.1 | 7692 | UTR 3 | XP_016859876.1 | |||
XM_017004388.1 | 7692 | UTR 3 | XP_016859877.1 | |||
XM_017004389.1 | 7692 | UTR 3 | XP_016859878.1 | |||
XM_017004390.1 | 7692 | UTR 3 | XP_016859879.1 | |||
XM_017004391.1 | 7692 | UTR 3 | XP_016859880.1 |