Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGTTGAGATGCGAAGGGCGTT[C/T]AAAGAAAACCTGGAGACTTCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP35 - Rho GTPase activating protein 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004491.4 | 1308 | Silent Mutation | TTC,TTT | F,F 436 | NP_004482.4 | |
XM_011526873.2 | 1308 | Silent Mutation | TTC,TTT | F,F 436 | XP_011525175.1 | |
XM_017026714.1 | 1308 | Silent Mutation | TTC,TTT | F,F 436 | XP_016882203.1 |