Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGACCATCTGGTGTCAGGGGAC[C/T]CTGGAGGCTCGGGAGTACCGTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604821 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRB4 - leukocyte immunoglobulin like receptor B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278426.3 | 395 | Silent Mutation | ACC,ACT | T,T 52 | NP_001265355.2 | |
NM_001278427.3 | 395 | Silent Mutation | ACC,ACT | T,T 52 | NP_001265356.2 | |
NM_001278428.3 | 395 | Silent Mutation | ACC,ACT | T,T 52 | NP_001265357.2 | |
NM_001278429.3 | 395 | Silent Mutation | ACC,ACT | T,T 81 | NP_001265358.2 | |
NM_001278430.3 | 395 | Intron | NP_001265359.2 | |||
XM_017026215.1 | 395 | Silent Mutation | ACC,ACT | T,T 93 | XP_016881704.1 | |
XM_017026216.1 | 395 | Silent Mutation | ACC,ACT | T,T 93 | XP_016881705.1 | |
XM_017026217.1 | 395 | Silent Mutation | ACC,ACT | T,T 93 | XP_016881706.1 |