Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCTGGGGAAACCCAGGATCTC[C/T]GACTGCCTGGACATTTGCATTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159350 MIM: 610709 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCC - mutated in colorectal cancers | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085377.1 | 1209 | Intron | NP_001078846.1 | |||
NM_002387.2 | 1209 | Intron | NP_002378.1 | |||
XM_005271991.2 | 1209 | Intron | XP_005272048.1 | |||
XM_017009473.1 | 1209 | Intron | XP_016864962.1 | |||
XM_017009474.1 | 1209 | Intron | XP_016864963.1 |
TSSK1B - testis specific serine kinase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032028.3 | 1209 | Silent Mutation | TCA,TCG | S,S 339 | NP_114417.1 |