Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATATTGAGAGGGTAAGCTCATTG[C/T]TGAGATCAAACACTCGTTTCTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611372 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAP1 - small ArfGAP 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001044305.2 | Intron | NP_001037770.1 | ||||
NM_001281439.1 | Intron | NP_001268368.1 | ||||
NM_001281440.1 | Intron | NP_001268369.1 | ||||
NM_021940.4 | Intron | NP_068759.2 | ||||
XM_005248760.4 | Intron | XP_005248817.1 | ||||
XM_011536045.2 | Intron | XP_011534347.1 | ||||
XM_011536046.2 | Intron | XP_011534348.1 | ||||
XM_011536047.2 | Intron | XP_011534349.1 | ||||
XM_011536048.2 | Intron | XP_011534350.1 | ||||
XM_017011170.1 | Intron | XP_016866659.1 |