Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCCTGCCCAGGGCTGCCCAGAG[A/T]TAGCCAACGCGTCCCTGTGCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610223 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RIN3 - Ras and Rab interactor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319987.1 | Intron | NP_001306916.1 | ||||
NM_024832.4 | Intron | NP_079108.3 | ||||
XM_011537164.2 | Intron | XP_011535466.1 | ||||
XM_011537165.2 | Intron | XP_011535467.1 | ||||
XM_017021651.1 | Intron | XP_016877140.1 | ||||
XM_017021652.1 | Intron | XP_016877141.1 | ||||
XM_017021653.1 | Intron | XP_016877142.1 | ||||
XM_017021654.1 | Intron | XP_016877143.1 |
Set Membership: |
HapMap |