Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615566 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BTBD3 - BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282550.1 | Intron | NP_001269479.1 | ||||
NM_001282551.1 | Intron | NP_001269480.1 | ||||
NM_001282552.2 | Intron | NP_001269481.1 | ||||
NM_001282554.1 | Intron | NP_001269483.1 | ||||
NM_014962.3 | Intron | NP_055777.1 | ||||
NM_181443.3 | Intron | NP_852108.2 | ||||
XM_017027728.1 | Intron | XP_016883217.1 | ||||
XM_017027729.1 | Intron | XP_016883218.1 | ||||
XM_017027730.1 | Intron | XP_016883219.1 |
Set Membership: |
Validated |