Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGCCTGACAGGTTGTGACTTG[C/G]GTAGAGAAGTCATGAAGGCCAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608199 MIM: 616561 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGLYRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PGLYRP2 - peptidoglycan recognition protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052890.3 | 2130 | Intron | NP_443122.3 | |||
XM_006722633.3 | 2130 | UTR 3 | XP_006722696.1 |
RASAL3 - RAS protein activator like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |