Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605953 | ||||||||||||||||||||
Literature Links: |
ASAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASAP1 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247996.1 | 5303 | UTR 3 | NP_001234925.1 | |||
NM_018482.3 | 5303 | UTR 3 | NP_060952.2 | |||
XM_005250925.1 | 5303 | UTR 3 | XP_005250982.1 | |||
XM_006716563.3 | 5303 | UTR 3 | XP_006716626.1 | |||
XM_006716564.1 | 5303 | UTR 3 | XP_006716627.1 | |||
XM_006716565.3 | 5303 | UTR 3 | XP_006716628.1 | |||
XM_006716566.1 | 5303 | UTR 3 | XP_006716629.1 | |||
XM_011517052.2 | 5303 | UTR 3 | XP_011515354.1 | |||
XM_011517053.1 | 5303 | UTR 3 | XP_011515355.1 | |||
XM_017013467.1 | 5303 | UTR 3 | XP_016868956.1 | |||
XM_017013468.1 | 5303 | UTR 3 | XP_016868957.1 | |||
XM_017013469.1 | 5303 | UTR 3 | XP_016868958.1 |