Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615210 MIM: 615423 | ||||||||||||||||||||
Literature Links: |
PCNP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCNP - PEST proteolytic signal containing nuclear protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320395.1 | Intron | NP_001307324.1 | ||||
NM_001320397.1 | Intron | NP_001307326.1 | ||||
NM_001320398.1 | Intron | NP_001307327.1 | ||||
NM_001320399.1 | Intron | NP_001307328.1 | ||||
NM_001320400.1 | Intron | NP_001307329.1 | ||||
NM_001320401.1 | Intron | NP_001307330.1 | ||||
NM_020357.2 | Intron | NP_065090.1 | ||||
XM_017006922.1 | Intron | XP_016862411.1 |
TRMT10C - tRNA methyltransferase 10C, mitochondrial RNase P subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |