Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTTTCAGAGGACAATCTTATG[C/T]CTTACTTTAATTTCTGGTGCTCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616348 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF670-ZNF695 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF670-ZNF695 - ZNF670-ZNF695 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF695 - zinc finger protein 695 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204221.1 | Intron | NP_001191150.1 | ||||
NM_020394.4 | Intron | NP_065127.4 |
Set Membership: |
HapMap |