Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACCCTGTCTTCTTCGCCTCGGC[A/G]CCGGTGCTCATCTCCATCCTGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300828 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTCHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTCHD1 - patched domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173495.2 | 101 | Silent Mutation | GCA,GCG | A,A 31 | NP_775766.2 | |
XM_011545449.2 | 101 | Silent Mutation | GCA,GCG | A,A 31 | XP_011543751.1 |
Set Membership: |
HapMap |