Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAGTTAATCAACTTTTCCAAAC[C/T]GCACAGTTAGTAAATGCCAGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CC2D2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CC2D2B - coiled-coil and C2 domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001732.3 | Intron | NP_001001732.2 | ||||
NM_001159747.1 | Intron | NP_001153219.1 | ||||
XM_011539789.2 | Intron | XP_011538091.1 | ||||
XM_011539790.2 | Intron | XP_011538092.1 | ||||
XM_011539791.2 | Intron | XP_011538093.1 | ||||
XM_011539792.2 | Intron | XP_011538094.1 | ||||
XM_011539793.2 | Intron | XP_011538095.1 | ||||
XM_011539795.2 | Intron | XP_011538097.1 | ||||
XM_017016240.1 | Intron | XP_016871729.1 |
ENTPD1-AS1 - ENTPD1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |