Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGACCAGGGGACTGGAGTGCGG[A/G]GCCGGCTGAAGGGCAAGAGGACAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGES3L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTGES3L - prostaglandin E synthase 3 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142653.1 | Intron | NP_001136125.1 | ||||
NM_001142654.1 | Intron | NP_001136126.1 | ||||
NM_001261430.1 | Intron | NP_001248359.1 |
PTGES3L-AARSD1 - PTGES3L-AARSD1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136042.2 | Intron | NP_001129514.2 | ||||
NM_025267.3 | Intron | NP_079543.1 |
RUNDC1 - RUN domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321381.1 | Intron | NP_001308310.1 | ||||
NM_173079.3 | Intron | NP_775102.2 | ||||
XM_005257078.3 | Intron | XP_005257135.1 | ||||
XM_005257080.3 | Intron | XP_005257137.1 |
Set Membership: |
HapMap |