Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300201 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYSLTR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
CHB (Han Chinese)
|
|||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
CYSLTR1 - cysteinyl leukotriene receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282186.1 | 1341 | Silent Mutation | NP_001269115.1 | |||
NM_001282187.1 | 1341 | Silent Mutation | NP_001269116.1 | |||
NM_001282188.1 | 1341 | Silent Mutation | NP_001269117.1 | |||
NM_006639.3 | 1341 | Silent Mutation | NP_006630.1 |
Set Membership: |
HapMap Validated |