Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602835 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GAS2 - growth arrest specific 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143830.1 | Intron | NP_001137302.1 | ||||
NM_005256.3 | Intron | NP_005247.1 | ||||
NM_177553.2 | Intron | NP_808221.1 | ||||
XM_011519971.2 | Intron | XP_011518273.1 | ||||
XM_011519972.2 | Intron | XP_011518274.1 | ||||
XM_011519975.1 | Intron | XP_011518277.1 | ||||
XM_011519978.2 | Intron | XP_011518280.1 | ||||
XM_017017528.1 | Intron | XP_016873017.1 | ||||
XM_017017529.1 | Intron | XP_016873018.1 | ||||
XM_017017530.1 | Intron | XP_016873019.1 | ||||
XM_017017531.1 | Intron | XP_016873020.1 | ||||
XM_017017532.1 | Intron | XP_016873021.1 |