Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTCTCTCTCTATCCTTCATAC[C/G]TGCCACAGAAACAAAGAGGACATGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Spock1 PubMed Links |
Spock1 - sparc/osteonectin, cwcv and kazal-like domains proteoglycan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166463.1 | Intron | NP_001159935.1 | ||||
NM_001166464.1 | Intron | NP_001159936.1 | ||||
NM_001166465.1 | Intron | NP_001159937.1 | ||||
NM_001166466.1 | Intron | NP_001159938.1 | ||||
NM_009262.3 | Intron | NP_033288.2 |