Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACATGGACAGCGTTTCTAGCCGC[A/G]CGGTCGGGAGCCGCTCCCTGAGGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Socs5 PubMed Links |
Socs5 - suppressor of cytokine signaling 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019654.2 | 953 | Missense Mutation | NP_062628.2 |