Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm21949 PubMed Links |
Gm21949 - predicted gene, 21949 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113419.2 | Intron | NP_001106890.1 |
Schip1 - schwannomin interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113420.1 | Intron | NP_001106891.1 | ||||
NM_001113421.1 | Intron | NP_001106892.1 | ||||
NM_001282044.1 | Intron | NP_001268973.1 | ||||
NM_001282045.1 | Intron | NP_001268974.1 | ||||
NM_013928.5 | Intron | NP_038956.2 |