Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAATTCCATGACAGGAGGGTTA[C/T]TGAATGTGTTTACTGTGCCATCCTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mctp1 PubMed Links |
Mctp1 - multiple C2 domains, transmembrane 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030174.2 | Intron | NP_084450.2 |