Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PIK3CA | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_006218.3 | NP_006209.2 | 5515965 | c.250G>A | p.D84N |
12579 | c.178C>A | p.Q60K | |||
6475681 | c.172C>T | p.L58F | |||
7347211 | c.273_274insACCGA | p.L92fs*10 | |||
5736834 | c.244delT | p.F83fs*17 | |||
6475604 | c.263G>T | p.R88L | |||
7347215 | c.289_290insTG | p.P97fs*4 | |||
3408448 | c.268T>G | p.C90G | |||
6573714 | c.213A>G | p.V71V | |||
6444823 | c.184C>A | p.L62I | |||
5347117 | c.225A>C | p.Q75H | |||
27493 | c.277C>T | p.R93W | |||
7346712 | c.286C>G | p.Q96E | |||
729832 | c.193G>A | p.E65K | |||
3737049 | c.253G>A | p.E85K | |||
6191573 | c.280C>T | p.L94F | |||
6191359 | c.278G>C | p.R93P | |||
3695922 | c.241G>T | p.E81* | |||
5346986 | c.161G>A | p.R54K | |||
5945917 | c.249T>G | p.F83L | |||
6191568 | c.274C>T | p.L92F | |||
5954758 | c.252T>C | p.D84D | |||
6163880 | c.260G>C | p.R87T | |||
5346971 | c.248T>C | p.F83S | |||
86041 | c.278G>A | p.R93Q | |||
5151687 | c.201T>C | p.S67S | |||
6913225 | c.247_249TTT>AAG | p.F83K | |||
6475679 | c.170C>A | p.P57H | |||
7450115 | c.275_276ins17 | p.R93fs*13 | |||
4653656 | c.211G>A | p.V71I | |||
7347340 | c.281_284delTTTT | p.F95fs*4 | |||
746 | c.263G>A | p.R88Q | |||
27502 | c.241G>A | p.E81K | |||
7347209 | c.272_273insT | p.L92fs*12 | |||
3408450 | c.269G>A | p.C90Y | |||
7347213 | c.277_278insCCATAGAGA | p.L92_R93insPIE | |||
3364898 | c.214A>G | p.S72G | |||
3732754 | c.155A>T | p.E52V | |||
6941830 | c.232G>A | p.E78K | |||
6920563 | c.168C>A | p.Y56* | |||
7412594 | c.277C>G | p.R93G | |||
3737048 | c.169C>T | p.P57S | |||
1753026 | c.269G>C | p.C90S | |||
7346040 | c.135delA | p.K46fs*26 | |||
6967333 | c.200C>T | p.S67F | |||
5347052 | c.242A>C | p.E81A | |||
6983144 | c.290C>A | p.P97Q | |||
1041454 | c.210C>T | p.F70F | |||
6915327 | c.197C>T | p.S66F | |||
304777 | c.248T>A | p.F83Y | |||
6475677 | c.132C>A | p.T44T | |||
163484 | c.238G>A | p.E80K | |||
6475767 | c.225A>G | p.Q75Q | |||
4832940 | c.268T>C | p.C90R | |||
6914463 | c.278G>T | p.R93L | |||
6746448 | c.224A>C | p.Q75P | |||
7115810 | c.197C>G | p.S66C | |||
6475616 | c.210C>A | p.F70L | |||
39166 | c.223C>G | p.Q75E | |||
XM_006713658.3 | XP_006713721.1 | - | - | - | |
XM_011512894.2 | XP_011511196.1 | - | - | - | |
GenBank | AB210020.1 | - | - | - | |
AK292940.1 | - | - | - | ||
AK302608.1 | - | - | - | ||
BC113601.1 | - | - | - | ||
BC113603.1 | - | - | - | ||
BX473473.1 | - | - | - | ||
BX484734.1 | - | - | - | ||
U79143.1 | AAB39753.1 | - | - | - | |
Z29090.1 | CAA82333.1 | - | - | - |
Literature Links: |
PIK3CA PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |