Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277074.1 | NP_001264003.1 | 7 | 516 | ||
NM_144716.5 | NP_653317.2 | 8 | 671 | |||
NR_102269.1 | 7 | 534 | ||||
XM_011533396.2 | XP_011531698.1 | 7 | 870 | |||
GenBank mRNA | AK096749.1 | - | 7 | 494 | ||
AK098120.1 | - | 1 | 2083 | |||
BC020830.1 | - | 7 | 497 | 4771328 | ||
BM803652.1 | - | 7 | 441 | |||
BX098833.1 | - | 5 | 295 |