Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMS22L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198468.2 | NP_940870.2 | 25 | 4174 | ||
XM_006715430.1 | XP_006715493.1 | 25 | 4105 | |||
XM_017010657.1 | XP_016866146.1 | 24 | 4092 | |||
GenBank mRNA | AK128060.1 | - | 25 | 4174 | ||
CR749603.1 | - | 5 | 784 |