Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019855.4 | NP_062829.1 | 6 | 1060 | ||
XM_017026992.1 | XP_016882481.1 | 8 | 1261 | |||
GenBank mRNA | AF169159.1 | - | 6 | 1053 |