Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TELO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016111.3 | NP_057195.2 | 2 | 407 | ||
XM_011522773.2 | XP_011521075.1 | 2 | 398 | |||
XM_011522774.2 | XP_011521076.1 | 2 | 596 | |||
XM_011522775.2 | XP_011521077.1 | 2 | 379 | |||
XM_011522776.2 | XP_011521078.1 | 2 | 466 | |||
XM_011522777.2 | XP_011521079.1 | 2 | 398 | |||
XM_011522778.2 | XP_011521080.1 | 2 | 398 | |||
XM_017023914.1 | XP_016879403.1 | 2 | 379 | |||
GenBank mRNA | AB014583.3 | - | 2 | 395 | ||
AK302103.1 | - | 2 | 145 | |||
AL080126.1 | - | 2 | 338 | |||
AL137394.1 | - | 2 | 338 | |||
BC017188.2 | - | 1 | 163 | 3139743 | ||
DB494123.1 | - | 2 | 372 |