Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005061.2 | NP_005052.1 | 8 | 1081 | ||
XM_011522571.2 | XP_011520873.1 | 8 | 1541 | |||
GenBank mRNA | AK313162.1 | - | 8 | 1093 | ||
BC050413.1 | - | 8 | 1073 | 6204847 | ||
U65581.1 | - | 8 | 1082 |