Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100112.1 | NP_001093582.1 | 20 | 2386 | ||
NM_017534.5 | NP_060004.3 | 20 | 2386 | |||
GenBank mRNA | AF111784.1 | - | 18 | 2258 | ||
AK299440.1 | - | 20 | 2197 | |||
AK300702.1 | - | 9 | 872 | |||
BC126409.1 | - | 19 | 2329 | 8992125 | ||
BX510904.2 | - | 20 | 2349 |