Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004886.3 | NP_004877.1 | 10, 11 | 1841 | ||
XM_006722950.3 | XP_006723013.1 | 9, 10 | 1967 | |||
XM_006722951.3 | XP_006723014.1 | 7, 8 | 1652 | |||
GenBank mRNA | AB021638.1 | - | 10, 11 | 1831 | ||
AB209603.1 | - | 3, 4 | 4040 | |||
AF029110.1 | - | 1, 2 | 356 | |||
BC008338.1 | - | 8, 9 | 1688 | |||
BC086306.1 | - | 9, 10 | 1700 | 6371716 |