Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008326.1 | NP_032352.1 | 3 | 551 | ||
GenBank mRNA | AK002545.1 | - | 3 | 573 | ||
AK167558.1 | - | 2 | 405 | |||
AK171743.1 | - | 3 | 532 | |||
BC145957.1 | - | 3 | 363 | 40131260 | ||
KC541561.1 | - | 1 | 58 | |||
U19119.1 | - | 3 | 551 |