Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290783.1 | NP_001277712.1 | 7 | 1537 | ||
NM_026137.5 | NP_080413.2 | 7 | 1571 | |||
NM_028542.1 | NP_082818.1 | 7 | 1823 | |||
GenBank mRNA | AF220146.1 | - | 6 | 1093 | ||
AF353243.1 | - | 7 | 1217 | |||
AF513713.1 | - | 7 | 1040 | |||
AK006837.1 | - | 6 | 1059 | |||
AK017549.1 | - | 7 | 1153 | |||
BC057366.1 | - | 7 | 1537 | |||
BC080853.1 | - | 7 | 1486 | |||
BC108332.1 | - | 7 | 1327 | 6493226 | ||
BC120553.1 | - | 7 | 1074 | 40129476 | ||
BC125287.1 | - | 7 | 1073 | 40129802 |