Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsb66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181694.2 | NP_859045.2 | 4 | 529 | ||
XM_006233762.1 | XP_006233824.1 | 4 | 692 | |||
XM_006233763.2 | XP_006233825.1 | 4 | 908 | |||
XM_006233764.2 | XP_006233826.1 | 4 | 1363 | |||
XM_006233766.1 | XP_006233828.1 | 4 | 692 | |||
GenBank mRNA | AY121839.1 | - | 3 | 479 | ||
BC097268.1 | - | 4 | 529 | 7457912 |