Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031803.1 | NP_113991.1 | 7, 8 | 797 | ||
GenBank mRNA | AF059273.1 | - | 7, 8 | 797 | ||
AF205778.1 | - | 691 | ||||
AF205779.1 | - | 691 | ||||
AF205780.1 | - | 691 | ||||
FQ211504.1 | - | 841 |