Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 4, 5 | 707 | ||
NM_001291696.1 | NP_001278625.1 | 3, 4 | 407 | |||
NM_012312.4 | NP_036444.1 | 4, 5 | 707 | |||
GenBank mRNA | AF285438.1 | - | 3, 4 | 632 | ||
AJ002102.1 | - | 4, 5 | 669 | |||
AJ002103.1 | - | 4, 5 | 672 | |||
AM409094.1 | - | 4, 5 | 673 | |||
AY366243.1 | - | 4, 5 | 682 | |||
AY366244.1 | - | 4, 5 | 671 | |||
BC108917.1 | - | 4, 5 | 674 | 40017299 | ||
BC119631.1 | - | 4, 5 | 691 | 40115568 | ||
BC119632.1 | - | 4, 5 | 691 | 40115570 | ||
BC125146.1 | - | 3, 4 | 656 | |||
BC126856.1 | - | 4, 5 | 674 | 40017301 | ||
FR750994.1 | - | 4, 5 | 707 | |||
L41347.1 | - | 4, 5 | 691 | |||
L76667.1 | - | 4, 5 | 389 | |||
L76668.1 | - | 3, 4 | 383 | |||
U24079.1 | - | 4, 5 | 670 | |||
X89893.1 | - | 4, 5 | 672 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 4, 5 | 710 | ||
XM_017026782.1 | XP_016882271.1 | 4, 5 | 688 | |||
XM_017026783.1 | XP_016882272.1 | 4, 5 | 1315 | |||
GenBank mRNA | AF022045.1 | - | 4, 5 | 679 | ||
AF285431.1 | - | 4, 5 | 705 | |||
AF285432.1 | - | 3, 4 | 632 | |||
AY366237.1 | - | 4, 5 | 708 | |||
AY366238.1 | - | 4, 5 | 654 | |||
AY366239.1 | - | 4, 5 | 702 | |||
AY366240.1 | - | 4, 5 | 710 | |||
AY366241.1 | - | 4, 5 | 709 | |||
AY388471.1 | - | 4, 5 | 702 | |||
AY789055.1 | - | 5, 6 | 705 | |||
AY789056.1 | - | 4, 5 | 672 | |||
BC069344.1 | - | 4, 5 | 705 | 7262296 | ||
KJ699235.1 | - | 4, 5 | 945 | |||
L41267.1 | - | 5, 6 | 704 | |||
U24076.1 | - | 5, 6 | 705 | |||
U24078.1 | - | 4, 5 | 712 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 4, 5 | 701 | ||
GenBank mRNA | AF285434.1 | - | 3, 4 | 632 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 4, 5 | 709 | ||
GenBank mRNA | AF022048.1 | - | 4, 5 | 672 | ||
AF285435.1 | - | 3, 4 | 632 | |||
AY789057.1 | - | 4, 5 | 672 | |||
BC032422.1 | - | 4, 5 | 685 | 5220339 | ||
BC050730.1 | - | 4, 5 | 703 | 5219336 | ||
BC110613.1 | - | 4, 5 | 686 | 40031619 | ||
FN395076.1 | - | 4, 5 | 672 | |||
FN552550.1 | - | 4, 5 | 654 | |||
L41268.1 | - | 4, 5 | 709 | |||
L76662.1 | - | 4, 5 | 683 | |||
L76663.1 | - | 4, 5 | 683 | |||
U24074.1 | - | 4, 5 | 693 | |||
U50546.1 | - | 4, 5 | 389 | |||
U73395.1 | - | 4, 5 | 693 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 4, 5 | 685 | ||
GenBank mRNA | AF022046.1 | - | 4, 5 | 685 | ||
AF285437.1 | - | 3, 4 | 632 | |||
AM999888.1 | - | 4, 5 | 680 | |||
X89892.1 | - | 4, 5 | 692 | |||
X98858.1 | - | 4, 5 | 672 |
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 4, 5 | 683 | ||
GenBank mRNA | AF022047.1 | - | 4, 5 | 676 | ||
AM851002.1 | - | 4, 5 | 708 | |||
AY789063.2 | - | 3, 4 | 656 | |||
L76670.1 | - | 4, 5 | 683 | |||
X97231.1 | - | 4, 5 | 731 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 4, 5 | 739 | ||
NM_012314.5 | NP_036446.3 | 5, 6 | 761 | |||
GenBank mRNA | AF002255.1 | - | 5, 6 | 718 | ||
AJ417554.1 | - | 3, 4 | 630 | |||
AJ417555.1 | - | 3, 4 | 652 | |||
AJ620615.2 | - | 3, 4 | 623 | |||
AJ620616.2 | - | 3, 4 | 623 | |||
AY102624.1 | - | 2, 3 | 362 | |||
AY366245.1 | - | 4, 5 | 706 | |||
AY366246.1 | - | 4, 5 | 657 | |||
AY388472.1 | - | 4, 5 | 710 | |||
AY789064.2 | - | 4, 5 | 634 | |||
BC096694.1 | - | 4, 5 | 683 | 40026979 | ||
BC101977.1 | - | 4, 5 | 683 | 40004312 | ||
BC103693.1 | - | 4, 5 | 683 | 40004314 | ||
BC108918.1 | - | 5, 6 | 674 | 40017300 | ||
BC125147.1 | - | 3, 4 | 656 | |||
BC127084.1 | - | 4, 5 | 689 | 40120468 | ||
BC144566.1 | - | 4, 5 | 761 | 9053094 | ||
BC171788.1 | - | 4, 5 | 761 | 9054442 | ||
L76671.1 | - | 4, 5 | 683 | |||
U24077.1 | - | 4, 5 | 663 | |||
X94609.1 | - | 4, 5 | 727 |