Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304388.1 | NP_001291317.1 | 8 | 2704 | ||
GenBank mRNA | AK093463.1 | - | 9 | 1725 | ||
AK128822.1 | - | 1393 | ||||
BC129820.1 | - | 9 | 1044 | 40113145 | ||
BC129821.1 | - | 8 | 1640 |