Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012444.2 | NP_036576.1 | 13 | 1249 | ||
NM_198265.1 | NP_937998.1 | 12 | 1135 | |||
XM_005260379.3 | XP_005260436.1 | 14 | 1300 | |||
XM_005260380.3 | XP_005260437.1 | 14 | 1294 | |||
XM_005260381.3 | XP_005260438.1 | 13 | 1261 | |||
XM_011528756.2 | XP_011527058.1 | 13 | 1186 | |||
XM_011528757.2 | XP_011527059.1 | 14 | 1134 | |||
GenBank mRNA | AF149310.1 | - | 13 | 1189 | ||
AF169385.1 | - | 13 | 1264 | |||
BC033591.1 | - | 12 | 1097 | 5167214 |