Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153696.2 | NP_710163.1 | 1, 2 | 439 | ||
XM_017017319.1 | XP_016872808.1 | 6, 7 | 792 | |||
GenBank mRNA | AF261715.1 | - | 1, 2 | 439 |