Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145028.1 | NP_001138500.1 | 3 | 257 | ||
XM_005259891.4 | XP_005259948.1 | 4 | 458 | |||
XM_011527965.2 | XP_011526267.1 | 3 | 378 | |||
GenBank mRNA | AW130597.1 | - | 1 | 235 | ||
CR747190.1 | - | 2 | 380 |