Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001541.3 | NP_001532.1 | 2 | 755 | ||
GenBank mRNA | AB096250.1 | - | 2 | 1249 | ||
D89617.1 | - | 2 | 730 | |||
S67070.1 | - | 1 | 400 |