Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131035.1 | NP_001124507.1 | 2 | 207 | ||
NM_001131036.1 | NP_001124508.1 | 2 | 179 | |||
NM_032280.2 | NP_115656.1 | 2 | 564 | |||
GenBank mRNA | AK314160.1 | - | 2 | 507 | ||
AL512712.1 | - | 1 | 3228 | |||
BC032736.1 | - | 2 | 144 | 5540043 | ||
BG776648.1 | - | 2 | 207 | |||
CB126982.1 | - | 2 | 164 | |||
CN261947.1 | - | 2 | 134 |