Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080997.2 | NP_001074466.1 | 3 | 348 | ||
NM_001270440.1 | NP_001257369.1 | 4 | 621 | |||
NM_001270441.1 | NP_001257370.1 | 3 | 301 | |||
NM_001270442.1 | NP_001257371.1 | 3 | 289 | |||
NM_001270443.1 | NP_001257372.1 | 3 | 277 | |||
NM_001270444.1 | NP_001257373.1 | 3 | 265 | |||
NM_001270445.1 | NP_001257374.1 | 2, 3 | 259 | |||
NM_031429.2 | NP_113617.1 | 4 | 621 | |||
XM_005260089.1 | XP_005260146.1 | 4 | 696 | |||
XM_005260090.3 | XP_005260147.1 | 3 | 559 | |||
XM_011528343.1 | XP_011526645.1 | 3 | 362 | |||
XM_011528344.2 | XP_011526646.1 | 3 | 412 | |||
XM_011528345.1 | XP_011526647.1 | 3 | 357 | |||
XM_017027343.1 | XP_016882832.1 | 4 | 651 | |||
XM_017027344.1 | XP_016882833.1 | 3, 4 | 603 | |||
GenBank mRNA | AB593122.1 | - | 3 | 301 | ||
AK093672.1 | - | 4 | 621 | |||
AY028917.1 | - | 3 | 325 | |||
BC005063.1 | - | 4 | 609 | 3344818 | ||
BG471112.1 | - | 3 | 262 | |||
BM688006.1 | - | 3 | 253 | |||
BM688127.1 | - | 3 | 254 | |||
BM708463.1 | - | 3 | 348 | |||
BQ639040.1 | - | 3, 4 | 223 | |||
EL946853.1 | - | 3 | 281 |