Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000835.4 | NP_000826.2 | 3 | 629 | ||
NM_001278553.1 | NP_001265482.1 | 3 | 883 | |||
NR_103735.1 | 3 | 629 | ||||
XM_006721845.3 | XP_006721908.1 | 2 | 1895 | |||
XM_006721846.3 | XP_006721909.1 | 2 | 1896 | |||
XM_011524686.2 | XP_011522988.1 | 2 | 1900 | |||
XM_011524687.2 | XP_011522989.1 | 2 | 1897 | |||
XM_011524688.2 | XP_011522990.1 | 2 | 1896 | |||
XM_011524689.2 | XP_011522991.1 | 3 | 620 | |||
XM_011524692.2 | XP_011522994.1 | 2 | 1893 | |||
XM_017024544.1 | XP_016880033.1 | 2 | 1898 | |||
GenBank mRNA | AB208799.1 | - | 2 | 2702 | ||
BC041128.1 | - | 3 | 575 | |||
BC140801.1 | - | 3 | 507 | 9021669 | ||
L76224.1 | - | 2 | 473 | |||
U77782.1 | - | 3 | 661 |