Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081249.1 | NP_001074718.1 | 8 | 5716 | ||
NM_019389.2 | NP_062262.2 | 7 | 2836 | |||
GenBank mRNA | D16263.1 | - | 6 | 2402 | ||
D28599.1 | - | 8 | 5454 |